Learn More
Electron Microscopy Sciences mini-BUCK™ High Flow Calibrator, 120 V
Supplier:  Electron Microscopy Sciences 6524064
Encompass_Preferred
The mini-BUCK Primary Flow Calibrator is the industry standard for day-to-day flow verification and calibration of your personal and environmental sampling pumps. It is also suitable for verifying rotameters, setting instrument panels, and calibrating laboratory instruments. No more lugging oversized calibration panels. No more rotameter inaccuracies. The mini BUCK is accurate at any attitude and can be used for flow measurements with sampling media. With 1/2" port.
Flow: 100cc/min 30LPM
Catalog No. 50-339-58
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.