Learn More
Cole-Parmer Traceable Jumbo Refrigerator/Freezer Thermometer with Calibration; 1 Bottle Probe
Supplier: Cole-Parmer 9446074
Thermometer is designed for monitoring freezers, water baths, heating blocks, incubators, refrigerators and other general-purpose applications. Display simultaneously shows maximum, minimum, and current probe temperatures. Min/max monitors high/low readings for any time period. Alarm is programmable in 1° increments. The 10-ft (3 m) micro-cable with solid-state probe permits doors to close on it. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.