Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma Milli-Q™ SR 240L Purification System Kit

Pure RO water production station with two 3.5 L SQ Switch Tanks and SQ Gravity Valve

Supplier:  MilliporeSigma ZSR240RLTK

Catalog No. ZSR240RLTK


Explore all special offers
Add to Cart

Description

Description

  • The Milli-Q™ SR 240L water purification system delivers pure reverse osmosis (RO) water to two mobile SQ Switch 3.5 L tanks.
  • It can be equipped with a Gravity valve (optional) to dispense pure RO water directly from the SQ Switch tank base.
  • Kit requires 1 Power Cord, Cartridge, and Installation (not included).
Specifications

Specifications

2 x 3.5 L
≤42 L/hr. Make-up Flow Rate (Pure RO water), ≤6 L/min. Distribution Flow Rate (Pure RO water from SQ Gravity valve)
Water Purification System
Potable Tap Water
Reverse Osmosis (Type 3)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.