Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block for Microtubes 0.5 mm, 30 Wells
Supplier:  Electron Microscopy Sciences 6312426
Encompass_Preferred
These versatile soild aluminum blocks have an anondized coating that resists the harshest environments. Provide maximum heat retention. All blocks include thermometer well for easily measuring block temperature. Fit all our Modular Block Dri-Baths.
Tube size: 0.5 mm
Wells: 30
Catalog No. 50-322-53
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.