Learn More
Andwin Scientific MICROTUBE BRUSH 10/PK
SDP
Supplier:  Andwin Scientific 17030006
Encompass
The Nylon Semimicro Centrifuge Tube Brush provides effective and thorough cleaning of centrifuge tubes. This product has an overall length of 18 centimeters and has a tapered diameter from 1.3 centimeters to 5 millimeters. It is sold in a pack of 10.
Catalog No. NC9071231
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.