Learn More
Fisherbrand™ Microscope Objective, Finite Optical Systems
Microscope Objective works with all finite based optical systems
Supplier: Fisherbrand™ 03000084
Specifications
Achromat Objective | |
RMS Threading | |
All Finite Based Optical Systems | |
100X (Oil) |