Learn More
Electron Microscopy Sciences Microplate Genie, 50hz, British Plug, 230 V
Supplier: Electron Microscopy Sciences 61040BR
Microplate Genie features high speed 750-3200 RPM mixing speed for increased mixing efficiency in every well and uniform mixing in any microplate format. Heat will not be transferred to your microplates. The fan (located in the bottom of the unit) directs air downward and out of the unit. As a result, room air is constantly drawn into the unit underneath the tray that holds the microplates, insuring that samples will not be heated.
May be used to shake any single microplate.
Small vortexing orbit of 1.0 mm for thorough mixing regardless of sample viscosity.
Speed and orbit combine to offer true vortexing action in each well of a microplate.
Accepts most microplate types within the recommendations of the Society for Biomolecular Screening (SBS), even in 384-well formats.
Built-in timer operates from 1 - 60 minutes or continuous for hands-free use.
May be used in cold rooms and incubators for temperature sensitive applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.