Learn More
Andwin Scientific MICROPLATE ADHESIVE FILM -40C
Supplier: Andwin Scientific 29200000
The microplate film offer reduced evaporation, spillage, and contamination between wells. This product also features a secure seal, which removes "edge effects" in sensitive ELISA assays. The films can also seal tissue-culture plates for short-term storage, incubation, and containment of biohazards. These can work in a temperature range from -40C to 120C. Nonsterile and non-pierceable.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.