Learn More
Contec™ MicroGenesis Mop
BIOPHARMA
MicroGenesis Mop 100% synthetic, disposable microfiber mop pad effectively removes fine particles and large debris from floors, providing superior removal of visible contaminants. Designed to be used wet or dry for general facilities cleaning in ISO Class 8 environments and above. It is compatible with a wide range of disinfectants, including quats, peroxides, and bleach. These synthetic pads are designed to provide uniform surface coverage.
Supplier:  Contec™ MGMP0002
 View more versions of this product
Catalog No. 19-165-900
Description
Description
Features & BenefitsSynthetic construction is compatible with common disinfectants such as quats, peroxides and bleachNew pad for each use simplifies quality control, inventory and space management, all without cross-contamination riskCompatible with most existing hardware
Specifications
Specifications
Backer Plate
Non-sterile
Single-use
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.