Learn More
Andwin Scientific MICRO COVERGLASS 22X40 1 OZ
SDP
Supplier:  Andwin Scientific 7220403
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness. Size: 22x40mm; Thickness 1 (.16-.19 mm); Pieces 76
Catalog No. 50-948-954
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.