Learn More
Andwin Scientific MICRO COVERGLASS 22X22 1 OZ
SDP
Supplier:  Andwin Scientific 7220010
Encompass_Preferred
The micro cover glasses are made from clear white, borosilicate glass that provide superior clarity. This product has a size of 22 x 22 mm and thickness of 0.13-0.17 mm. It also features resistance against corrosion and a perfectly flat design. This is sold in a 1 ounce pack of 156 cover glasses.
Catalog No. 50-948-948
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.