Learn More
Andwin Scientific MICRO CLEANING SOLUTION 1GAL
SDP
Supplier:  Andwin Scientific 71056
Encompass_Preferred
The 1 gallon Micro Cleaning Solution is used specially for ultrasonic cleaning machines, glass, metals, plastics and rubber. It can remove radioactive contaminants and contains no phosphates.
Catalog No. 50-930-739
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.