Learn More
Electron Microscopy Sciences Micro Cleaning Solution 1 QT
Supplier: Electron Microscopy Sciences 71054
Specially designed for ultrasonic cleaning machines. Cleans glass, most metals, plastics, and rubber. You can work with any concentration. Contains no phosphates and it is biodegradable. Even removes radioactive contaminants. Not to be used on zinc, magnesium, or aluminum.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.