Learn More
Andwin Scientific MICRO-90 CLEANING SOL'N 12CS
SDP
Supplier:  Andwin Scientific 89210138
Encompass_Preferred
The Micro-90� Concentrated Alkaline Cleaning Solution is a powerful cleaning concentrate that can efficiently remove oil, grease, wax, tar, flux, particulates, hard water stains, and biological debris. This solution is composed of chelants, ionic, and non-ionic ingredients that generate many cleaning actions such as lifting, dispersing, emulsifying, sequestering, suspending, and decomposing. This product is ideal for laboratory cleaning, industrial applications, and critical cleaning processes. It is packaged in a 1 liter plastic bottle and is sold in a case of 12.
Catalog No. NC0233367
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.