Learn More
Andwin Scientific MICRO-90 CLEANING SOL 1LT
Supplier: Andwin Scientific 1810005
The 1-liter Micro-90 cleaning solution is an equipment cleaner that can remove most biological materials such as oil, grease, wax, flux, particulates or hard water stains. It is ideal for glass, most metals, plastic, rubber, fabric and ultrasonic cleaners. It is potent and easily rinses clean at any concentration.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.