Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mettler Toledo™ Plastic Syringe, for Mettler Toledo™ Titrators
For use with DL31 Karl Fischer titrator.
Supplier: Mettler Toledo™ 71482
Description
- Capacity: 10mL
- Quantity: 100/Pk.
Specifications
10 mL Plastic Syringe, Pack of 100 | |
Titration | |
10 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction