Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Methylene Blue, Certified, C.I. DcA-24 25 GR

Supplier:  Electron Microscopy Sciences 18600

Encompass_Preferred

(Basic Blue 9; Swiss Blue)
CAS #7220-79-3 C₁₆H₁₈N₃SCl •₃H₂O F.W. 373.89
Solubility: 9.5% Water; 6.0% Alcohol; 6.5% Cellosolve; 10.0% Glycol; 0.0% Xylene

Used to distinguish autonomic nerves.

Richardson, K.C., (1969) Anat. Rec., 164, 231. General stain for animals, histology. A simple rapid staining procedure for methacrylate embedded tissue sections using chromotrope 2R and Methylene Blue.

Dougherty, M.M., and King, J.S., (1984) Stain Tech., 59, 149. An established routine method for differential staining of epoxy-embedded tissue sections.

Schroeder, H.E., Rossinsky, K., and Muller, W., (1980) Microsc. Acta 83, 111. Used with Basic Fuchsin for stain epoxy-embedded tissue.

Sato, T. and Shamoto, M. (1973). A simple rapid polychrome stain for epoxy-embedded tissue. Stain Technol., 48:223.

Catalog No. 50-271-67


Explore all special offers
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.