Learn More
Electron Microscopy Sciences Methyl Cellulose, 400csp 100 GR
Supplier:  Electron Microscopy Sciences 18560
Encompass_Preferred
(Methylcel MC; Cellulose Methyl Ether)
Used with polyethylene glycol in the presence of acidic uranyl acetate to stain cryo sections.
Catalog No. 50-271-61
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.