Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-(3-pyridyl)propionate, 98%, Thermo Scientific Chemicals

Catalog No. AAL1459006
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84199-98-4
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD01075660
InChI Key FQRRQPZYIYAPAV-UHFFFAOYSA-N
Synonym methyl 3-3-pyridyl propionate, methyl 3-pyridin-3-yl propanoate, methyl 3-3-pyridyl propanoate, methyl 3-3-pyridinyl propanoate, 3-pyridinepropanoicacid, methyl ester, 3-3-pyridyl-propionic acid methyl ester
PubChem CID 10329646
IUPAC Name methyl 3-pyridin-3-ylpropanoate
SMILES COC(=O)CCC1=CN=CC=C1

Specifications

Boiling Point 134°C (12 mmHg)
Refractive Index 1.501
Quantity 5 g
Beilstein 132852
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material Methyl 3-(3-pyridyl)propionate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.