Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Metallothionein Monoclonal Antibody (1F5), APC, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA545895
Description
A 1:1,000 dilution of MA5-45895 was sufficient for detection of Metallothionein in 1 μg of purified Pseudomonas aeruginosa Metallothionein (PmtA)-GST tagged protein by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.∣Detects 36 kDa band corresponding to Pseudomonas aeruginosa Metallothionein.
Metallothioneins (MTs) are a family of low molecular weight, heavy metal-binding proteins characterized by a high cysteine content and lack of aromatic amino acids. MTs bind 7 to 12 heavy metal atoms per molecule of protein. They are ubiquitous in the animal and plant kingdoms and are also found in prokaryotes. In mammals, the cysteine residues are absolutely conserved and serve to coordinate heavy metal atoms such as zinc, cadmium, and copper via mercaptide linkages.Specifications
Metallothionein | |
Monoclonal | |
1 mg/mL | |
2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.4 | |
0 | |
Mt1, PA2140 | |
Recombinant full length Pseudomonas aeruginosa Metallothionein protein | |
100 μg | |
Primary | |
Bacteria | |
Liquid |
ELISA, Western Blot | |
1F5 | |
APC | |
PA2140 | |
CES 1; CES1; LOC100343802; LOC100630794; Metallothionein; metallothionein 1; metallothionein 1a; metallothionein 1A (functional); metallothionein 1S; Metallothionein 2; Metallothionein 2A; Metallothionein IA; Metallothionein II; metallothionein-1; Metallothionein-1A; metallothionein-1A-like; Metallothionein2; metallothionein-I; Metallothionein-IA; MGC32848; Mt; MT 1A; MT 2; MT 2A; MT IA; MT II; Mt1; MT-1; MT1A; MT-1A; MT1S; MT2; MT2A; MTC; MT-I; MT-IA; Probable pmtA | |
Mouse | |
Protein G | |
RUO | |
20220066, 880550 | |
4° C | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction