Learn More
Andwin Scientific Metal-Free Centrifuge Tubes, Polypropylene, Sterile, 15 mL, 500/cs
SDP
Supplier:  Andwin Scientific 89049172
Encompass_Preferred
These leak-proof centrifuge tubes are ideal for environmental uses such as water analysis and other applications wherein tubes have the potential to contaminate samples.
Catalog No. NC1355544
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.