Learn More
Sklar Instruments Forceps, Merit Tuttle Forceps, Sklar 7 in., Fenestrated, Straight, Serrated, Stainless Steel, EA.
Supplier: Sklar Instruments 97617
Forceps, Merit Tuttle Forceps, Sklar, 7 in., Fenestrated, Straight, Serrated, Stainless Steel, EA. Merit Tuttle Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. These forceps have long shafts which make them useful for working in areas such as the intestines. These forceps may also be known as Singley forceps. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted from Pakistan steel and come with a 10 year guarantee.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.