Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ Men's Navy Flame Resistant Flex Knit Henley

Catalog No. 17204392 Shop All Red Kap Industries Products
Click to view available options
Size:

Men's Navy Flame Resistant Flex Knit Henley has a casual, contemporary look with lightweight fabric for added comfort and range of movement, curved flatlock seams for optimal fit, and a unique front pocket design with pencil stall.

  • Three-button front placket for enhanced appearance
  • Contrasting pattern on sleeve for athletic look
  • CAT 2/2112 left sleeve label leaves no question that you are protected on the job site
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Home Wash
  • Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m2) Flame Resistant Jersey
  • Blend: Body: 93% Cotton/7% Spandex Elastane, Contrast Panels: 93% Cotton/7% Spandex Elastane
  • Collar: Rib
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size

X-Large

Gender

Men's

Material

Cotton, Spandex

Color

Navy

Neck Style

Henley

Sleeve Style

Long Sleeve

Construction

Knit

Closure Type

Button

Disposable

Reusable

No. of Pockets

1

Hazard Risk Category

Category 2

Arc Flash Rating

8.3

Certifications/Compliance

NFPA 2112, NFPA 70E

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.