Learn More
Eurofins Discoverx MEK1, unactive, 250 µg. (14-420M)
Supplier: Eurofins Discoverx 14420M
N-terminal GST-tagged and C-terminal 6xHis-tagged recombinant, human full length MEK1. For detailed product information, visit www.discoverx.com/proteins.