Learn More
Andwin Scientific MEGA-CASSETTES
Supplier: Andwin Scientific 25608844
The Tissue-Tek� Mega-Cassette� is made to be used for processing and embedding larger, thicker specimens, including bone, eye, and teeth. There are three textured surfaces that can be written on to offer users easy identification and organization. The cassette are able to work with solvents and decalcifying solutions. This measures 30 mm x 24 mm x 11 mm.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.