Learn More
Medium-Chain Triglycerides, NF, 200 L, Spectrum Chemical
Supplier: Spectrum Chemical Manufacturing Corporation M4200200LTBL
Description
Medium-Chain Triglycerides, NF consist of a mixture of triglycerides of saturated fatty acids, mainly of caprylic (octanoyl) acid and capric (decanoyl) acid. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Specifications
200 L |