Learn More
Anton Paar Measuring system CC26 - 26mm cup and bob measuring system for ViscoQC 100 & ViscoQC 300
Supplier:  Anton Paar 191726
Encompass_Preferred
Measuring system consisting of bob B-CC26 and cup C-CC26 for measurement according to ISO 3219 and DIN 53019-1. The stainless steel measuring bob is equipped with Toolmaster and magnetic coupling. A stainless steel hook connects the spindle body of the bob to the magnetic coupling. The cup consists of an anodized aluminum wall, a PEEK bottom plug and an FKM O-ring which seals the gap between wall and bottom.Requires holder for cup, either DIN Adapter item NC1818345 or PTD 80 item NC1818588Suitable for use with Anton Paar rotational viscometers ViscoQC 100 – L item NC1818559, ViscoQC 100 – R item NC1818567, ViscoQC 100 – H item NC1818569, ViscoQC 300 – L item NC1818572, ViscoQC 300 – R item NC1818576, ViscoQC 300 – H item NC1818578.
Catalog No. NC1818602
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.