Learn More
Electron Microscopy Sciences Mayer's Hematoxylin, 1L
Supplier: Electron Microscopy Sciences 2604306
Mayer's Hematoxylin is used as a counterstain for procedures such as immunohistochemistry or laser microdissection.