Learn More
Andwin Scientific MAT LID FOR 2ML DP/W NS 50PK
SDP
Supplier:  Andwin Scientific 40002016
Encompass_Preferred
The 96-well deep well microplates can work with both automated and robotic systems. This product is also compatible to use cell harvesters and many microplate centrifuge rotors. The plates are resistant against chemicals and are ideal for high-throughput screening applications, including drug discovery and diagnostic research. This product is also alphanumerically labeled that allows convenient and easy sample identification. Each round well has a volume capacity of 0.5 mL and a conical bottom. It is sold in a pack of 50.
Catalog No. NC9009743
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.