Learn More
Eurofins Discoverx MAPKAP Kinase 2, unactive, 250 µg. (14-349M)
Supplier:  Eurofins Discoverx 14349M
Encompass_Preferred
Recombinant human MAPKAP Kinase 2 (MAPKAP-K2), amino acids 46-400, A399G, containing an N-terminal GST tag. MAPKAP Kinase 2 has been implicated as a cell cycle checkpoint kinase, joining the ranks of Chk1 and Chk2 as critical regulators of the DNA damage response in mammalian cells in recent studies (Manke, et al, 2005, and Abraham, 2005). This research indicates that MAPKAP Kinase 2, a serine/threonine kinase activated by p38, phosphorylates CDC25 B/C, generating a binding site for 14-3-3 proteins. The checkpoint kinases Chk1 and Chk2 are targets for drug discovery efforts designed to overcome cell cycle arrest due to DNA damage induced by chemotherapeutic agents. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8782
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.