Learn More
Sino Biological Manual pipette tip -300μL, Transparent, Box packing
Supplier: Sino Biological PTM300YNN
S-ProScience Universal Pipette tips, made of polypropylene, are sterile and DNase/RNase free. 96 tips per Rack.Our universal filter tips are designed to fit Eppendorf and Gilson pipettes, offering compatibility with a variety of pipette brands.• High clarity, 100% virgin polystyrene.• Manufactured in a class 100,000 clean Rooms• Dnase, Rnase, protease and pyrogen free• Electron beam sterilization, without chemical residue• Universal fit• Outstanding volume precision with fine point design• Minimal residual liquid adherence
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.