Learn More
MilliporeSigma™ Supelco™ Manual Bubble Flowmeters
A simple and precise way to measure flow rates of gases
Supplier:  MilliporeSigma™ Supelco™ 20415
 View more versions of this product
Catalog No. 11-100-3766
Description
Description
  • The most common device for measuring a volumetric flow rate
  • This unit has a burette of appropriate volume fitted with rubber bulb at the bottom and a side arm
  • Air is passed into the burette and the bulb filled with soap solution is squeezed to create a bubble
  • The time taken for the bubble to travel through a specific volume is used to calculate the flow rate
Specifications
Specifications
Graduated, 100 mL Increments
1000 mL
Determination of Volumetric Flow Rates
Includes a Glass Flow Tube, 5/16 in. Tygon™ Tubing (30 in.), Squeeze Bulb, Collapsible Stand, and A Bottle of Liquid Soap Solution
Large
Bubble Flowmeter Starter Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.