Learn More
MilliporeSigma™ Supelco™ Manual Bubble Flowmeter, Capillary Version
Determines volumetric flow rates by measuring the time taken by the gas stream to move a soap bubble through a specific volume
Supplier:  MilliporeSigma™ Supelco™ 23762U
 View more versions of this product
Catalog No. 11-100-4520
Description
Description
  • A simple and precise way to measure flow rates of gases
  • Allows the true split ratio to be determined by measuring actual flow rate
  • The flow tube is graduated in 0.01 mL increments to accomodate low flows that are typical of capillary GC applications
  • Two magnetic clamps hold the flow tube together so that the unit can be attached to any metal surface for easy storage
  • A version that includes a sampling stand is offered for added portability
Specifications
Specifications
Without Stand
Measuring Gas Flow Rates
0.5 mL Bubble Flowmeter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.