Learn More
STEMCELL Technologies Easy 250 EasySep™ Magnet, For isolating: 1.25 x 10^10 cells
Supplier: STEMCELL Technologies 1000821
Scale up your manual cell isolations from large-volume samples, such as leukopaks and whole blood. Rather than splitting your cell suspension and performing multiple rounds of isolations, with the Easy 250 EasySep™ magnet you can process up to 225 mL and 1.25 x 1010 cells in a single isolation.Use the Easy 250 EasySep™ magnet with a standard T-75 cm2 cell culture flask and EasySep™ reagents to separate cells from large-volume samples without the use of columns. With its strong, high-gradient internal magnetic field, the magnet isolates cells labeled with magnetic EasySep™ particles, allowing you to simply pipette out the unlabeled cell fraction and obtain cells that are immediately ready for downstream applications. Save time and obtain highly purified cells with EasySep™ column-free technology. Learn more about how EasySep™ technology works or get an overview of our other magnets on our EasySep™ Cell Separation Magnets page.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.