Learn More
Julabo USA Inc MAGIO MS-1200F R1270 EA 230V
SDP
Supplier:  Julabo USA Inc 9032717.N1.33
Encompass_Preferred
Refrigerated/heating circulator, JULABO, MAGIO MS-1200F 230V/60Hz, R1270 Refrigerant; -50…+200 °C; 2kW heater. Cooling: 1.26kW at 20C; 1.11kW at 0C; 640W at -20C, 390W @ -30C, 220W at -40C. Adjustable pressure/suction pump 16-31L/min, 3.5-13.3PSI. Color touchscreen UI with USB, RS232/RS485, Ethernet, MODBUS communication, USB data-logging; Pt100 port, multi-lingual, programmable. M16x1 pump fittings 8 and 12mm barbs. 5-7.5L bath tank. 13 x 18.5 x 27.6 in, 97lbs
Catalog No. NC2751045
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.