Learn More
TELEFLEX LLC MAD Nasal Device with 1 mL Syringe and Vial Adapter, 25/pk
Supplier:  TELEFLEX LLC MAD130
Encompass
The MAD Nasal™ Intranasal Mucosal Atomization Device from Teleflex is the safe and painless way to deliver medications. Atomizes in any position. The soft, conical plug on the tip forms a seal with the nostril, preventing expulsion of fluid. The spray atomizes drugs into a fine mist of particles 30-100 microns in size.
Catalog No. NC1677709
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.