Learn More
New England Biolabs, Inc. M13mp18 Single-stranded DNA – 10 µg
Supplier: New England Biolabs, Inc. N4040S
The single-stranded viral DNA is isolated from M13mp18. M13mp18 is a M13 lac phage vector which contains single HindIII, SphI, SbfI, PstI, SalI (AccI/ HincII), XbaI, BamHI, SmaI (XmaI), KpnI (Acc65I), SacI and EcoRI sites within the gene encoding Galactosidase. This DNA preparation is useful as a standard and has been tested as a template in the dideoxy-nucleotide termination method of sequencing DNA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.