Learn More
Teledyne ISCO, Inc M1 Class Single Piston Pump; Eccentric Drive; 0.0 - 100.0 Ml/Min.; 250 Psi
Supplier:  Teledyne ISCO, Inc M1100SRX
Encompass
M1 Class Single Piston Pump; Eccentric Drive; 0.0 - 100.0 mL/min.; 250 psi; Stainless Steel Fluid Path; without Piston Wash; without Pulse Dampener; Standard Seal (stainless steel energizer); Fitting package for 1/8"""" tubing
Catalog No. 50-211-9954
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.