Learn More
Cytiva M-FC Broth, Fecal Coliforms, 2 mL glass (20/pkg)
Supplier:  Cytiva 68100
Encompass_Preferred
Sterile Ampoule Media Pre-Measured for Maximum Efficiency and Convenience-Meets Membrane Filter (MF) Technique Standard Method requirements with 2 mL ampoules*-Maximizes efficiency with premixed and presterilized media-Choose between plastic or glass ampoules for your applications-Simplify pouring of the media with wide-mouth glass ampoules-All plastic ampoule media is economically packaged with 50 ampoules per box This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3350
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.