Learn More
Hach Company m-ColiBlue24 Broth, 100mL Bottle (50 test)
Supplier: Hach Company 2608442
In 24 hours, Hach's m-ColiBlue24 Broth give you simultaneous results for total coliforms and E. coli. Hach's ready-to-use media eliminates measuring, mixing, and autoclaving steps necessary to prepare dehydrated media. Glass ampules (26084-20), plastic ampules (260845-00), 100mL bottles (26084-42) of m-ColiBlue24 and agar plates (28052-15), afford maximum shelf life and ease of use for pre-measured and prepared media. Enumerate total coliforms and E. coli on one petri dish. Differentiate colonies easily - red and blue indicate total coliforms and blue specifies E. coli. Use m-ColiBlue24 Broth to accurately monitor and evaluate total coliform and E. coli in drinking water; wastewater; bottled water; beverages; surface, ground, and well water; and ultrapure, chemical processing and pharmaceutical processing waters.m-ColiBlue24 should only be used with pads manufactured by the following companies/brands: Pall Corporation, Sartorius, and Millipore EO.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.