Learn More
Medchemexpress LLC HY-113123 5mg Medchemexpress, LysoPC(14:0/0:0) CAS:20559-16-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1131235MG
Encompass_Preferred
Medchemexpress, HY-113123 5mg LysoPC(14:0/0:0) CAS:20559-16-4 LysoPC(14:0/0:0) is a lysophospholipid (LyP). It is a monoglycerophospholipid in which a phosphorylcholine moiety occupies a glycerol substitution site. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9227
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.