Learn More
Enzo Life Sciences LYSO-ID Red detection kit (GFP-CERTIFIED) (100tests)
Supplier: Enzo Life Sciences ENZ510050100
LYSO-ID Red Lysosomal Detection Kit (GFP-CERTIFIED) is an ideal choice for fluorescence co-localization imaging with GFP-tagged proteins, providing spectrally pure red signal. By contrast, many red-emitting lysosomal probes photoconvert to green fluorescent species or display metachromatic artifacts wherein emission occurs both in the red and green regions of the spectrum, leading to spurious results in GFP co-localization experiments [Freundt et al (2007) Cell Res. 17(11): 956- 958; Nadrigny et al (2007) Biophys J. 93(3):969-980]. The LYSO-ID Red Lysosomal Detection Kit (GFP-CERTIFIED)contains a proprietary acidic organelle-selective dye suitable for staining live cells. The dye is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes, as well as High Content Screening (HCS) platforms. Applications: Fluorescence microscopy, Fluorescent detection .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.