Learn More
Enzo Life Sciences LYSO-ID Green detection kit (2x96wells)
Supplier: Enzo Life Sciences ENZ510340100
The LYSO-ID Green detection kit contains an acidic organelle-selective dye suitable for live-cell staining. Conventional fluorescent stains for acidic organelles, such as Acridine Orange (ENZ-52405), form metachromatic artifacts that interfere with multicolor imaging applications. LYSO-ID Green dye generates emission profiles that can be multiplexed with other fluorophores. The dye accumulates in acidic compartments, such as endosomes, lysosomes, and secretory vesicles. Low micromolar concentrations of LYSO-ID Green dye are sufficient for staining mammalian cells. This has been validated with the human cervical carcinoma cell line, HeLa, the human T-lymphocyte cell line, Jurkat, and the human bone osteosarcoma epithelial cell line, U2OS. The LYSO-ID Green dye is a new green-emitting, cell-permeable small organic probe molecule that spontaneously localizes to live cell acidic organelles. Applications: Fluorescence microscopy, Fluorescent detection .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.