Learn More
New England Biolabs, Inc. LyoPrime Luna Probe One-Step RT-qPCR Mix with UDG - 96 rxns
Supplier: New England Biolabs, Inc. L4001P
The LyoPrime Luna Probe One-Step RT-qPCR Mix with UDG is offered in a lyophilized format, allowing for ambient/room temperature shipping and storage prior to use. The mix contains all necessary components for one-step RT-qPCR except primers, probe(s), and RNA sample. It can be reconstituted by simply adding water to a 2-4X concentration, allowing for increased sample input. Performance in multiplexing applications has been optimized, with sensitive, linear detection achieved for up to 5 targets across a range of inputs. It is available in 2 x 60 reaction glass vials or a 96-well plate (1 reaction per well).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.