Learn More
Electron Microscopy Sciences EMS LYNX Tissue Processor Process Accelerator (LPA)
Supplier:  Electron Microscopy Sciences LX12668
Encompass_Preferred
The Lynx-LPA paraffin vacuum infiltration systems was designed to allow a greater through-put of tisssue specimens without compromising the critical infiltration step. Once your initial tissues reach the first paraffin step you can transfer the cassette holder to the Lynx-Pal for vacuum infiltration while you un-burden the Lynx to start another process cycle. The LPA has 3 positions to accept three HP cassette holder which conveniently uses the same Aluminum vial as the Lynx processor to contain paraffin. Each Aluminum vial is fitted with a platform to allow the stir-bar to circulate the melted wax.
Catalog No. 50-192-6635
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.