Learn More
Eurofins Discoverx Luteinizing Hormone, 10 µg. (92-1112)
Supplier: Eurofins Discoverx 921112
Luteinizing hormone (LH) is a pituitary reproductive hormone which acts in concert with follicle stimulating hormone. It regulates menstrual cycle by triggering ovulation and corpus luteum development in women. Also triggers production of testosterone by stimulating leydig cells in men. Acts as an agonist for LHCGR. Provided as 10 μg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter, PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.