Learn More
New England Biolabs, Inc. LunaScript RT SuperMix Kit – 25 reactions
Supplier: New England Biolabs, Inc. E3010S
LunaScript RT SuperMix Kit features an optimized master mix containing all the necessary components for first strand cDNA synthesis in the context of a two-step RT-qPCR workflow. It includes the thermostable Luna Reverse Transcriptase, which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is also included to protect template RNA from degradation. The LunaScript RT SuperMix contains random hexamer and poly-dT primers, allowing even coverage across the length of the RNA targets. In addition, the LunaScript RT SuperMix Kit contains a blue dye, providing a visual indicator that can be followed throughout the two-step RT-qPCR process. The LunaScript RT SuperMix offers robust, linear, and sensitive detection using total RNA inputs as high as 1 µg and as low as single copies of RNA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.