Learn More
New England Biolabs, Inc. Luna Probe One-Step RT-qPCR 4X Mix with UDG (No ROX) 500 reactions
SDP
Supplier:  New England Biolabs, Inc. M3029L
Encompass
The Luna Probe One-Step RT-qPCR Mix with UDG (No ROX) is supplied at a 4X concentration and enables higher amounts of sample input which is relevant for applications where RNA present in low abundance is of interest such as pathogen detection. The mix consolidates the components for one-step RT-qPCR into a single tube including Luna WarmStart RT Hot Start Taq DNA Polymerase dNTPs and Murine RNase Inhibitor in an optimized buffer. Combining Hot Start Taq DNA Polymerase with a novel WarmStart-activated reverse transcriptase allows for dual control of enzyme activity via reversible aptamer-based inhibition. This temperature-dependent activation helps to prevent undesirable non-specific priming and extension prior to thermocycling providing added security for setting up reactions at room temperature. This formulation contains no reference dye and is compatible with any instrument that does not require ROX (if ROX normalization is needed ROX can be added.)
Catalog No. 50-217-2586
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.