Learn More
New England Biolabs, Inc. Luna Cell Ready Lysis Module – 100 reactions
Supplier: New England Biolabs, Inc. E3032S
Cell cultures are often analyzed for gene expression or treatment responses as a proxy for a living organism. Traditionally, RNA is extracted and purified from treated cells via column-based or chemical methods. The Luna Cell Ready Lysis Module is a fast, convenient, and sensitive alternative for evaluating RNA expression levels without performing traditional RNA extraction steps. The module coordinates cell lysis, RNA release, and genomic DNA removal simultaneously, resulting in cell lysate ready for direct RT-qPCR analysis with Luna Universal One-Step RT-qPCR kits. Similar to other Luna products, the lysis buffer includes an inert blue tracking dye for visual assistance throughout the workflow.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.