Learn More
New England Biolabs, Inc. Luna Probe One-Step RT-qPCR 4X Mix with UDG - 200 rxns
SDP
Supplier:  New England Biolabs, Inc. M3019S
Encompass
The Luna Probe One-Step RT-qPCR 4X Mix with UDG is designed for real-time detection of target RNA sequences using hydrolysis probes. In a single tube, RNA is first converted to cDNA by a reverse transcriptase, then a DNA-dependent DNA polymerase amplifies the cDNA, enabling quantitation via real time or quantitative PCR (qPCR). Probe-based qPCR/RT-qPCR monitors an increase in fluorescence upon 5' to 3' exonuclease cleavage of a quenched, target-specific probe to measure DNA amplification at each cycle of a PCR. At a point where the fluorescence signal is confidently detected over the background fluorescence, a quantification cycle or Cq value can be determined. Cq values can be used to evaluate relative target abundance between two or more samples.
Catalog No. 50-202-3994
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.